Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1-Chloropentane, 98%
Click to view available options
Quantity:
50g
250g
1000g
Specifications
Refractive Index | 1.4125 |
Density | 0.881 |
Flash Point | 3°C (37°F) |
Boiling Point | 106°C to 108°C |
Melting Point | -99°C |
Quantity | 1000g |
Odor | Sweet |
Linear Formula | CH3(CH2)4Cl |
UN Number | UN1107 |
Merck Index | 14,606 |
Show More |
Safety and Handling
GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P330-P363-P370+P378q-P501c
H225-H302+H312+H332
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES
EINECSNumber : 208-846-4
TSCA : Yes
Recommended Storage : Ambient temperatures