Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Chloropentane, 98%

Click to view available options
Quantity:
50g
250g
1000g
543-59-9
C5H11Cl
106.593
MFCD00001015
SQCZQTSHSZLZIQ-UHFFFAOYSA-N
pentyl chloride,pentane, 1-chloro,n-amyl chloride,n-pentyl chloride,amyl chloride,chloropentane,unii-0eg9msd3nk,0eg9msd3nk,pentane, chloro,n-butylcarbonyl chloride
10977
1-chloropentane
CCCCCCl
543-59-9
C5H11Cl
106.593
MFCD00001015
SQCZQTSHSZLZIQ-UHFFFAOYSA-N
pentyl chloride,pentane, 1-chloro,n-amyl chloride,n-pentyl chloride,amyl chloride,chloropentane,unii-0eg9msd3nk,0eg9msd3nk,pentane, chloro,n-butylcarbonyl chloride
10977
1-chloropentane
CCCCCCl
Boiling Point 106°C to 108°C
Quantity 50g
Odor Sweet
Flash Point 3°C (37°F)
Melting Point -99°C
Linear Formula CH3(CH2)4Cl
UN Number UN1107
Merck Index 14,606
Formula Weight 106.6
Percent Purity 98%
Chemical Name or Material 1-Chloropentane
Show More Show Less

GHS H Statement
H225-H302-H312-H332
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
P273-P403
Avoid release to the environment.
Store in a well-ventilated place.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES

TSCA : TSCA