Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholinocarbenium hexafluorophosphate, 98%

Manufacturer:  Alfa Aesar™ H3374622

Quantity 100g

 View more versions of this product

Catalog No. AAH3374622


 

Due to product restrictions, please to purchase or view availability for this product.


Specifications

Description

Applications
(1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholinocarbenium hexafluorophosphate is used as a coupling reagent in the peptide synthesis. It is suitable for solution phase & solid phase peptide synthesis, visual or colorimetric reaction monitoring possible.

Solubility
Soluble in DMF 1mmol in 2mL.

Notes
Air and moisture sensitive. Store away from air, heat, water/ moisture and oxidizing agents.
Description & Specifications

Chemical Identifiers

1075198-30-9
428.27
COMU
C12H19F6N4O4P
MFCD11975052
(1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholinocarbenium hexafluorophosphate

Specifications

100g
C12H19F6N4O4P
MFCD11975052
≈160°C
98%
1075198-30-9
428.27
COMU
(1-Cyano-2-ethoxy-2-oxoethylidenaminooxy)dimethylamino-morpholinocarbenium hexafluorophosphate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit