Alfa Aesar™ 1-Cyclohexene-1-boronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H5103603

Quantity 1g

 View more versions of this product

Catalog No. AAH5103603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141091-37-4
    Molecular Formula C12H21BO2
    Formula Weight 208.1
    MDL Number MFCD05663845
    Chemical Name or Material 1-Cyclohexene-1-boronic acid pinacol ester

    Specifications

    Formula Weight 208.1
    Molecular Formula C12H21BO2
    MDL Number MFCD05663845
    Flash Point 103°C (217°F)
    Quantity 1g
    CAS 141091-37-4
    Recommended Storage 2° to 8°C
    Chemical Name or Material 1-Cyclohexene-1-boronic acid pinacol ester
    Assay 95%

    2° to 8°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit