Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexylpiperazine 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
17766-28-8
C10H20N2
168.28
MFCD00044809
XPDSXKIDJNKIQY-UHFFFAOYSA-N
n-cyclohexylpiperazine,1-cyclohexyl-piperazine,cyclohexylpiperazine,piperazine, 1-cyclohexyl,1-cyclohexylpiperazin,n-cyclohexyl piperazine,1-cyclohexyl piperazine,piperazine,1-cyclohexyl,4-1-cyclohexyl piperazine,acmc-1c47v
87298
1-cyclohexylpiperazine
C1CCC(CC1)N2CCNCC2
17766-28-8
C10H20N2
168.28
MFCD00044809
XPDSXKIDJNKIQY-UHFFFAOYSA-N
n-cyclohexylpiperazine,1-cyclohexyl-piperazine,cyclohexylpiperazine,piperazine, 1-cyclohexyl,1-cyclohexylpiperazin,n-cyclohexyl piperazine,1-cyclohexyl piperazine,piperazine,1-cyclohexyl,4-1-cyclohexyl piperazine,acmc-1c47v
87298
1-cyclohexylpiperazine
C1CCC(CC1)N2CCNCC2
Infrared Spectrum Authentic
Melting Point 28.0°C to 32.0°C
Quantity 25g
Packaging Glass bottle
Refractive Index 1.6011
Color Brown-Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 168.28
Physical Form Low Melting Solid
Percent Purity 98%
Chemical Name or Material 1-Cyclohexylpiperazine, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Immediately call a POISON CENTER or doctor/physician.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 241-750-0

TSCA : TSCA