Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclopenteneacetonitrile, 90%, ACROS Organics™

Click to view available options
Quantity:
10g
Packaging:
Glass bottle
22734-04-9
C7H9N
107.156
MFCD00001398
LEWVRAMNXUWSFL-UHFFFAOYSA-N
1-cyclopentenylacetonitrile,1-cyanomethyl cyclopentene,2-cyclopent-1-en-1-yl acetonitrile,cyclopent-1-ene-1-acetonitrile,1-cyclopentylacetonitrile,1-cyclopenteneacetonitrile,1-cyclopentene-1-acetonitrile,1-cyclopentenylacetronitrile,2-cyclopenten-1-yl acetonitrile
89811
2-(cyclopenten-1-yl)acetonitrile
C1CC=C(C1)CC#N
22734-04-9
C7H9N
107.156
MFCD00001398
LEWVRAMNXUWSFL-UHFFFAOYSA-N
1-cyclopentenylacetonitrile,1-cyanomethyl cyclopentene,2-cyclopent-1-en-1-yl acetonitrile,cyclopent-1-ene-1-acetonitrile,1-cyclopentylacetonitrile,1-cyclopenteneacetonitrile,1-cyclopentene-1-acetonitrile,1-cyclopentenylacetronitrile,2-cyclopenten-1-yl acetonitrile
89811
2-(cyclopenten-1-yl)acetonitrile
C1CC=C(C1)CC#N
Quantity 10g
Density 0.9510g/mL
Packaging Glass bottle
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula C5H7CH2CN
Formula Weight 107.15
Chemical Name or Material 1-Cyclopenteneacetonitrile

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning