Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(Cyclopropylmethyl)piperazine dihydrochloride, 97%

Click to view available options
Quantity:
1g
5g
373608-42-5
C8H18Cl2N2
213.146
MFCD08544420
XOERBUIHFQNXBA-UHFFFAOYSA-N
1-cyclopropylmethyl piperazine dihydrochloride,1-cyclopropylmethylpiperazine dihydrochloride,1-cyclopropylmethyl-piperazine dihydrochloride,c8h16n2.2clh,1-cyclopropylmethyl piperazine 2hcl,1-cyclopropylmethyl piperazine hydrochloride;piperazine,1-cyclopropylmethyl piperazine-hydrogen chloride 1/2
18403316
1-(cyclopropylmethyl)piperazine;dihydrochloride
C1CC1CN2CCNCC2.Cl.Cl
373608-42-5
C8H18Cl2N2
213.146
MFCD08544420
XOERBUIHFQNXBA-UHFFFAOYSA-N
1-cyclopropylmethyl piperazine dihydrochloride,1-cyclopropylmethylpiperazine dihydrochloride,1-cyclopropylmethyl-piperazine dihydrochloride,c8h16n2.2clh,1-cyclopropylmethyl piperazine 2hcl,1-cyclopropylmethyl piperazine hydrochloride;piperazine,1-cyclopropylmethyl piperazine-hydrogen chloride 1/2
18403316
1-(cyclopropylmethyl)piperazine;dihydrochloride
C1CC1CN2CCNCC2.Cl.Cl
Quantity 5g
Beilstein 6420036
Formula Weight 213.15
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(Cyclopropylmethyl)piperazine dihydrochloride