Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decanesulfonic acid, sodium salt, 96%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
13419-61-9
C10H21NaO3S
244.32
MFCD00007526
AIMUHNZKNFEZSN-UHFFFAOYSA-M
sodium decane-1-sulfonate,sodium 1-decanesulfonate,1-decanesulfonic acid sodium salt,1-decanesulfonic acid, sodium salt,decyl sodium sulfonate,sodium decane-1-sulphonate,ipc-alks-10,n-decylsulfonic acid, sodium salt,1-decanesulphonic acid, sodium salt,2-decanesulfonic acid, sodium salt
2724181
sodium;decane-1-sulfonate
CCCCCCCCCCS(=O)(=O)[O-].[Na+]
13419-61-9
C10H21NaO3S
244.32
MFCD00007526
AIMUHNZKNFEZSN-UHFFFAOYSA-M
sodium decane-1-sulfonate,sodium 1-decanesulfonate,1-decanesulfonic acid sodium salt,1-decanesulfonic acid, sodium salt,decyl sodium sulfonate,sodium decane-1-sulphonate,ipc-alks-10,n-decylsulfonic acid, sodium salt,1-decanesulphonic acid, sodium salt,2-decanesulfonic acid, sodium salt
2724181
sodium;decane-1-sulfonate
CCCCCCCCCCS(=O)(=O)[O-].[Na+]
Melting Point >300°C
Quantity 25g
Color White to Cream
Infrared Spectrum Authentic
Packaging Glass bottle
Loss on Drying 5% max. (100°C, 3 hrs) (vacuum)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula CH3(CH2)9SO3Na
Beilstein 04, III, 27
Solubility Information Solubility in water: soluble
Formula Weight 244.32
Physical Form Crystalline Powder
Percent Purity ≥95% (on dry substance)
Chemical Name or Material 1-Decanesulfonic acid, sodium salt, 96%
Show More Show Less