Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decanesulfonic Acid Sodium Salt, Approx. 99%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
13419-61-9
13419-61-9
Packaging Poly Pail
Quantity 1 kg
CAS Max % 1
Percent Purity Approx. 99%
Grade Reagent

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Safety Data Sheet (SDS) prior to use.