Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog No. AAL0664218
Click to view available options
Quantity:
50 g

1-Decanesulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Decanesulfonyl chloride is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61652-81-1
Molecular Formula C10H21ClO2S
Molecular Weight (g/mol) 240.79
MDL Number MFCD00051746
InChI Key WCKKWABZCCKDTN-UHFFFAOYSA-N
Synonym 1-decanesulfonyl chloride, decanesulfonylchloride, decanesulfonyl chloride, decylsulfonyl chloride, 1-decanesulphonylchloride, acmc-1b9lu
PubChem CID 109107
IUPAC Name decane-1-sulfonyl chloride
SMILES CCCCCCCCCCS(Cl)(=O)=O

Specifications

Melting Point 30°C to 34°C
Quantity 50 g
UN Number UN3261
Beilstein 1771852
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 240.79
Percent Purity 98%
Chemical Name or Material 1-Decanesulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.