Alfa Aesar™ 1-Decylboronic acid pinacol ester, 98%

Manufacturer: Alfa Aesar™ H3040314

Quantity 25g

 View more versions of this product

Catalog No. AAH3040314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 141091-38-5
    Molecular Formula C16H33BO2
    Formula Weight 268.25
    MDL Number MFCD12546186
    Synonym 2-(n-Decyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material 1-Decylboronic acid pinacol ester

    Specifications

    Formula Weight 268.25
    Molecular Formula C16H33BO2
    MDL Number MFCD12546186
    CAS 141091-38-5
    Quantity 25g
    Synonym 2-(n-Decyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
    Chemical Name or Material 1-Decylboronic acid pinacol ester
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit