Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Deoxy-1-nitro-L-iditol hemihydrate, 99%

Click to view available options
Quantity:
100mg
500mg
207226-23-1
C6H15NO8
229.185
MFCD00191972
OHFQXMNDKUHFDU-SIQASLMSSA-N
1-deoxy-1-nitro-l-iditol hemihydrate,2s,3r,4r,5s-6-nitrohexane-1,2,3,4,5-pentol hydrate
73995060
(2S,3R,4R,5S)-6-nitrohexane-1,2,3,4,5-pentol;hydrate
C(C(C(C(C(CO)O)O)O)O)[N+](=O)[O-].O
207226-23-1
C6H15NO8
229.185
MFCD00191972
OHFQXMNDKUHFDU-SIQASLMSSA-N
1-deoxy-1-nitro-l-iditol hemihydrate,2s,3r,4r,5s-6-nitrohexane-1,2,3,4,5-pentol hydrate
73995060
(2S,3R,4R,5S)-6-nitrohexane-1,2,3,4,5-pentol;hydrate
C(C(C(C(C(CO)O)O)O)O)[N+](=O)[O-].O
Melting Point 87°C to 89°C
Quantity 500mg
Formula Weight 220.19 (211.18 Anhydrous)
Percent Purity 99%
Chemical Name or Material 1-Deoxy-1-nitro-L-iditol hemihydrate