Learn More
Tocris Bioscience™ 1-Deoxymannojirimycin hydrochloride
α-Mannosidase I inhibitor
Supplier: Tocris Bioscience™ 1259/25
This item is not returnable.
View return policy
Description
1-Deoxymannojirimycin hydrochloride is a selective inhibitor of α-mannosidase I.Specifications
(2R,3R,4R,5R)-2-(Hydroxymethyl)-3,4,5-piperidinetriol hydrochloride | |
25 mg | |
C6H13NO4.HCl |
73465-43-7 | |
Soluble to 50mM in water | |
Observed MW: 199.63 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature