Learn More
Selleck Chemical LLC 1-Deoxynojirimycin S3839-5mg
SDP
Supplier:  Selleck Chemical LLC S38395MG
Encompass_Preferred
1-Deoxynojirimycin (duvoglustat moranolin) is a potent -glucosidase inhibitor and most commonly found in mulberry leaves It has therapeutic potency against diabetes mellitus
Catalog No. 50-310-9641