Alfa Aesar™ 1-Difluoromethoxy-3-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ B2226706

Quantity 5g

 View more versions of this product

Catalog No. AAB2226706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 22236-07-3
    Molecular Formula C7H5F2NO3
    Formula Weight 189.12
    MDL Number MFCD03407974
    Chemical Name or Material 1-Difluoromethoxy-3-nitrobenzene

    Specifications

    Formula Weight 189.12
    Odor Characteristic
    Quantity 5g
    Molecular Formula C7H5F2NO3
    MDL Number MFCD03407974
    CAS 22236-07-3
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point >110°C (230°F)
    Boiling Point 240° to 242°C
    Chemical Name or Material 1-Difluoromethoxy-3-nitrobenzene
    Assay 98%

    GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit