Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Difluoromethoxy-4-nitrobenzene, 98%

Click to view available options
Quantity:
5g
25g
1544-86-1
C7H5F2NO3
189.118
MFCD00085009
SVGGBARCOQPYMV-UHFFFAOYSA-N
4-difluoromethoxy nitrobenzene,1-difluoromethoxy-4-nitrobenzene,1-difluoromethoxy-4-nitro-benzene,4-difluoromethoxy-1-nitrobenzene,acmc-209dar,4-difluoromethoxynitrobenzene,svggbarcoqpymv-uhfffaoysa,4-nitro-alpha,alpha-difluoroanisole
2737014
1-(difluoromethoxy)-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])OC(F)F
1544-86-1
C7H5F2NO3
189.118
MFCD00085009
SVGGBARCOQPYMV-UHFFFAOYSA-N
4-difluoromethoxy nitrobenzene,1-difluoromethoxy-4-nitrobenzene,1-difluoromethoxy-4-nitro-benzene,4-difluoromethoxy-1-nitrobenzene,acmc-209dar,4-difluoromethoxynitrobenzene,svggbarcoqpymv-uhfffaoysa,4-nitro-alpha,alpha-difluoroanisole
2737014
1-(difluoromethoxy)-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])OC(F)F
Boiling Point 108°C to 110°C (1.5mmHg)
Odor Characteristic
Melting Point 33°C to 35°C
Quantity 5g
Formula Weight 189.12
Percent Purity 98%
Chemical Name or Material 1-Difluoromethoxy-4-nitrobenzene

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger