Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dimethylamino-2-nitroethylene, 98%, Thermo Scientific Chemicals

Catalog Number p-7081597
Click to view available options
Quantity:
1 g
5 g
1190-92-7
C4H8N2O2
116.12
MFCD00051519
JKOVQYWMFZTKMX-ONEGZZNKSA-N
1-dimethylamino-2-nitroethylene, e-n,n-dimethyl-2-nitroethenamine, dimethyl e-2-nitroethenyl amine, n,n-dimethyl-2-nitroethenamine, 1-dimethylamino-2-nitroethene, dimethyl 2-nitroethenyl amine, n,n-dimethyl-2-nitroethylenamine, n,n-dimethyl-2-nitroethyleneamine, n,n-dimethyl-n-2-nitrovinyl amine
637928
(E)-N,N-dimethyl-2-nitroethenamine
CN(C)C=C[N+](=O)[O-]
1190-92-7
C4H8N2O2
116.12
MFCD00051519
JKOVQYWMFZTKMX-ONEGZZNKSA-N
1-dimethylamino-2-nitroethylene, e-n,n-dimethyl-2-nitroethenamine, dimethyl e-2-nitroethenyl amine, n,n-dimethyl-2-nitroethenamine, 1-dimethylamino-2-nitroethene, dimethyl 2-nitroethenyl amine, n,n-dimethyl-2-nitroethylenamine, n,n-dimethyl-2-nitroethyleneamine, n,n-dimethyl-n-2-nitrovinyl amine
637928
(E)-N,N-dimethyl-2-nitroethenamine
CN(C)C=C[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 101°C to 105°C
Quantity 1 g
Beilstein 1903136
Formula Weight 116.12
Percent Purity 98%
Chemical Name or Material 1-Dimethylamino-2-nitroethylene
Hazard Category H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit