Alfa Aesar™ 1-Dimethylamino-2-propanol, 99%

Manufacturer: Alfa Aesar™ 43727K3

Quantity 2L

 View more versions of this product

Catalog No. AA43727K3

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 108-16-7
    Molecular Formula (CH3)2NCH2CH(OH)CH3
    Formula Weight 103.16
    MDL Number MFCD00004532
    UN Number UN2733
    Chemical Name or Material 1-Dimethylamino-2-propanol

    Specifications

    Formula Weight 103.16
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Refractive Index 1.4193
    Boiling Point 121° to 127°C
    UN Number UN2733
    Health Hazard 2 GHS H Statement
    H314-H318-H226-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.
    TSCA TSCA
    CAS 108-16-7
    Quantity 2L
    Health Hazard 1 Danger
    Density 0.83g/mL
    Molecular Formula (CH3)2NCH2CH(OH)CH3
    Physical Form Liquid
    Melting Point −40°C
    MDL Number MFCD00004532
    Flash Point 35°C (95°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    Chemical Name or Material 1-Dimethylamino-2-propanol
    Assay 99%

    GHS H Statement
    H314-H318-H226-H302
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Harmful if swallowed.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit