Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dimethylamino-2-propanol 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
Packaging:
Glass bottle
108-16-7
C5H13NO
103.16
MFCD00004532
NCXUNZWLEYGQAH-UHFFFAOYSA-N
1-dimethylamino-2-propanol,dimepranol,1-dimethylamino propan-2-ol,n,n-dimethylisopropanolamine,dimethyl 2-hydroxypropyl amine,2-propanol, 1-dimethylamino,dimethylisopropanolamine,dimepranol inn,n,n-dimethylamino-2-propanol
37511
1-(dimethylamino)propan-2-ol
CC(CN(C)C)O
108-16-7
C5H13NO
103.16
MFCD00004532
NCXUNZWLEYGQAH-UHFFFAOYSA-N
1-dimethylamino-2-propanol,dimepranol,1-dimethylamino propan-2-ol,n,n-dimethylisopropanolamine,dimethyl 2-hydroxypropyl amine,2-propanol, 1-dimethylamino,dimethylisopropanolamine,dimepranol inn,n,n-dimethylamino-2-propanol
37511
1-(dimethylamino)propan-2-ol
CC(CN(C)C)O
Viscosity 4.9 mPa.s (50°C)
Density 0.8500g/mL
Color Colorless to Yellow
Quantity 250g
Melting Point -85.0°C
Water 0.5% max.
Packaging Glass bottle
Refractive Index 1.4183 to 1.4203
Specific Gravity 0.85
Flash Point 26°C
Infrared Spectrum Authentic
Boiling Point 126.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2NCH2CH(OH)CH3
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in most organic solvents
Formula Weight 103.16
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Dimethylamino-2-propanol, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-556-4

RTECSNumber : UB3150000

TSCA : TSCA

Recommended Storage : Product may turn yellow on prolonged storage