Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt, 95%

Manufacturer:  Alfa Aesar™ B2212409

Quantity 10g

 View more versions of this product

Catalog No. AAB2212409


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

136997-71-2
258.29
1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt
C10H14N2O4S
MFCD00185917

Specifications

136997-71-2
≈180°C (dec.)
MFCD00185917
1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt
C10H14N2O4S
258.29
10g
95%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280g-P262
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.

Warning

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit