Alfa Aesar™ 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt, 95%

Manufacturer: Alfa Aesar™ B2212430

Quantity 250g

 View more versions of this product

Catalog No. AAB2212430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 136997-71-2
    Molecular Formula C10H14N2O4S
    Formula Weight 258.29
    MDL Number MFCD00185917
    Chemical Name or Material 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt

    Specifications

    Formula Weight 258.29
    TSCA TSCA
    Melting Point ≈180°C (dec.)
    Molecular Formula C10H14N2O4S
    MDL Number MFCD00185917
    CAS 136997-71-2
    Quantity 250g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280g-P262
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    Do not get in eyes, on skin, or on clothing.
    Chemical Name or Material 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt
    Assay 95%

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280g-P262
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves.
    Do not get in eyes, on skin, or on clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit