Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt, 95%

Click to view available options
Quantity:
10g
50g
250g
136997-71-2
C10H14N2O4S
258.292
MFCD00185917
SJHHXGGXZKPIDE-UHFFFAOYSA-N
1-n,n-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt,1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium inner salt,2-1-dimethylcarbamoyl pyridin-1-ium-4-yl ethanesulfonate,1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt,1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridin-1-ium,2-1-n,n-dimethylcarbamoyl-4-pyridyl ethanesulfonic acid,1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium betaine,1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridinium,2-1-dimethylcarbamoyl-4-pyridinio ethylsulfonate
3853422
2-[1-(dimethylcarbamoyl)pyridin-1-ium-4-yl]ethanesulfonate
CN(C)C(=O)[N+]1=CC=C(C=C1)CCS(=O)(=O)[O-]
136997-71-2
C10H14N2O4S
258.292
MFCD00185917
SJHHXGGXZKPIDE-UHFFFAOYSA-N
1-n,n-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt,1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium inner salt,2-1-dimethylcarbamoyl pyridin-1-ium-4-yl ethanesulfonate,1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium hydroxide inner salt,1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridin-1-ium,2-1-n,n-dimethylcarbamoyl-4-pyridyl ethanesulfonic acid,1-dimethylcarbamoyl-4-2-sulfoethyl pyridinium betaine,1-dimethylcarbamoyl-4-2-sulfonatoethyl pyridinium,2-1-dimethylcarbamoyl-4-pyridinio ethylsulfonate
3853422
2-[1-(dimethylcarbamoyl)pyridin-1-ium-4-yl]ethanesulfonate
CN(C)C(=O)[N+]1=CC=C(C=C1)CCS(=O)(=O)[O-]
Melting Point ~180°C (decomposition)
Quantity 50g
Formula Weight 258.29
Percent Purity 95%
Chemical Name or Material 1-Dimethylcarbamoyl-4-(2-sulfoethyl)pyridinium inner salt

GHS H Statement
H317
May cause an allergic skin reaction.

GHS P Statement
P261-P280g-P262
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
Do not get in eyes, on skin, or on clothing.

Warning

TSCA : TSCA