Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Diphenylmethyl)piperazine 97%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
Packaging:
Plastic bottle
841-77-0
C17H20N2
252.36
MFCD00038379
NWVNXDKZIQLBNM-UHFFFAOYSA-N
1-diphenylmethyl piperazine,benzhydrylpiperazine,n-benzhydrylpiperazine,1-benzhydryl-piperazine,diphenylmethylpiperazine,norcyclizine,normethylcyclizine,piperazine, 1-diphenylmethyl,4-benzhydrylpiperazine,n-diphenylmethyl piperazine
70048
1-benzhydrylpiperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3
841-77-0
C17H20N2
252.36
MFCD00038379
NWVNXDKZIQLBNM-UHFFFAOYSA-N
1-diphenylmethyl piperazine,benzhydrylpiperazine,n-benzhydrylpiperazine,1-benzhydryl-piperazine,diphenylmethylpiperazine,norcyclizine,normethylcyclizine,piperazine, 1-diphenylmethyl,4-benzhydrylpiperazine,n-diphenylmethyl piperazine
70048
1-benzhydrylpiperazine
C1CN(CCN1)C(C2=CC=CC=C2)C3=CC=CC=C3
Infrared Spectrum Authentic
Color White to Yellow
Packaging Plastic bottle
Flash Point 115°C
Water 2% max. (K.F.)
Quantity 500g
Melting Point 88.0°C to 93.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Solubility Information Solubility in water: 0.45g/L (20°C). Other solubilities: 305g/L acetone - >500g/L methanol, >500g/L ethanol - 215g/L 2-propanol, 346g/L dmf - >500g/L methyleenchloride
Formula Weight 252.36
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1-(Diphenylmethyl)piperazine, 97%
Show More Show Less

GHS P Statement Harmful if swallowed.

GHS P Statement IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

AIR SENSITIVE

EINECSNumber : 212-667-7

RTECSNumber : TL6465000

TSCA : TSCA