Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Docosanol, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Plastic bottle
661-19-8
C22H46O
326.61
MFCD00002939
NOPFSRXAKWQILS-UHFFFAOYSA-N
1-docosanol,behenyl alcohol,docosanol,behenic alcohol,docosyl alcohol,n-docosanol,abreva,tadenan,lidavol,lanette 22
12620
CHEBI:31000
docosan-1-ol
CCCCCCCCCCCCCCCCCCCCCCO
661-19-8
C22H46O
326.61
MFCD00002939
NOPFSRXAKWQILS-UHFFFAOYSA-N
1-docosanol,behenyl alcohol,docosanol,behenic alcohol,docosyl alcohol,n-docosanol,abreva,tadenan,lidavol,lanette 22
12620
CHEBI:31000
docosan-1-ol
CCCCCCCCCCCCCCCCCCCCCCO
Viscosity 7.5 mPa.s (80°C)
Flash Point 227°C
Color White
Melting Point 68.0°C to 72.0°C
Quantity 100g
Infrared Spectrum Authentic
Packaging Plastic bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)21OH
Beilstein 01, 431
Merck Index 15, 3444
Solubility Information Solubility in water: insoluble.
Formula Weight 326.61
Physical Form Pellets or Tablets
Percent Purity 98%
Chemical Name or Material 1-Docosanol, 98%
Show More Show Less

EINECSNumber : 211-546-6