Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecanesulfonic acid, sodium salt, 99+%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
2386-53-0
C12H25NaO3S
272.38
MFCD00007527
DAJSVUQLFFJUSX-UHFFFAOYSA-M
sodium 1-dodecanesulfonate,sodium dodecane-1-sulfonate,1-dodecanesulfonic acid sodium salt,sodium laurylsulfonate,1-dodecanesulfonic acid, sodium salt,unii-4gy1kjw8si,sodium dodecyl sulfonate,4gy1kjw8si,laurylsulfonic acid sodium salt,1-dodecanesulfonic acid, sodium salt 1:1
23665726
sodium;dodecane-1-sulfonate
CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
2386-53-0
C12H25NaO3S
272.38
MFCD00007527
DAJSVUQLFFJUSX-UHFFFAOYSA-M
sodium 1-dodecanesulfonate,sodium dodecane-1-sulfonate,1-dodecanesulfonic acid sodium salt,sodium laurylsulfonate,1-dodecanesulfonic acid, sodium salt,unii-4gy1kjw8si,sodium dodecyl sulfonate,4gy1kjw8si,laurylsulfonic acid sodium salt,1-dodecanesulfonic acid, sodium salt 1:1
23665726
sodium;dodecane-1-sulfonate
CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
Quantity 1g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Cream
Melting Point >300.0°C
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula CH3(CH2)11SO3Na
Solubility Information Solubility in water: soluble.
Formula Weight 272.38
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material 1-Dodecanesulfonic acid, sodium salt, 99+%
Show More Show Less