Alfa Aesar™ 1-Dodecyl isothiocyanate, tech. 90%

Manufacturer: Alfa Aesar™ B2180203

Quantity 1g

 View more versions of this product

Catalog No. AAB2180203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1072-32-8
    Molecular Formula CH3(CH2)11NCS
    Formula Weight 227.41
    MDL Number MFCD00041121
    UN Number UN1760
    Beilstein 1774889
    Synonym Lauryl isothiocyanate
    Chemical Name or Material 1-Dodecyl isothiocyanate
    Grade Technical Grade

    Specifications

    Formula Weight 227.41
    Odor Irritating
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    CAS 1072-32-8
    Beilstein 1774889
    Health Hazard 1 Danger
    Boiling Point 173° to 174°C (1mmHg)
    Health Hazard 2 GHS H Statement
    H301-H311-H314-H318-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if inhaled.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Molecular Formula CH3(CH2)11NCS
    MDL Number MFCD00041121
    UN Number UN1760
    Synonym Lauryl isothiocyanate
    Chemical Name or Material 1-Dodecyl isothiocyanate
    Assay 90%
    Grade Technical Grade

    GHS H Statement
    H301-H311-H314-H318-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if inhaled.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit