Alfa Aesar™ 1-Dodecylboronic acid pinacol ester, 98%

Manufacturer: Alfa Aesar™ H3118603

Quantity 1g

 View more versions of this product

Catalog No. AAH3118603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 177035-82-4
    Molecular Formula C18H37BO2
    Formula Weight 296.31
    MDL Number MFCD12546195
    Synonym 2-n-Dodecyl-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
    Chemical Name or Material 1-Dodecylboronic acid pinacol ester

    Specifications

    Formula Weight 296.31
    CAS 177035-82-4
    Boiling Point 123°C (0.2mmHg)
    Quantity 1g
    Molecular Formula C18H37BO2
    MDL Number MFCD12546195
    Synonym 2-n-Dodecyl-4,4,5,5-tetramethyl[1,3,2]dioxaborolane
    Chemical Name or Material 1-Dodecylboronic acid pinacol ester
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit