Alfa Aesar™ 1-Dodecylmagnesium bromide, 0.5M in MeTHF

Manufacturer: Alfa Aesar™ H51172AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH51172AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15890-72-9
    Molecular Formula C12H25BrMg
    Linear Formula CH3(CH2)11MgBr
    Formula Weight 273.54
    MDL Number MFCD00075165
    UN Number UN2924
    Chemical Name or Material 1-Dodecylmagnesium bromide

    Specifications

    Formula Weight 273.54
    Molecular Formula C12H25BrMg
    Linear Formula CH3(CH2)11MgBr
    MDL Number MFCD00075165
    CAS 15890-72-9
    UN Number UN2924
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
    Concentration 0.5M in MeTHF
    Quantity 500mL
    Chemical Name or Material 1-Dodecylmagnesium bromide

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapor.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P280-P305+P351+P338-P309-P310-P402
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    Store in a dry place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit