Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Eicosanol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
629-96-9
C20H42O
298.54
MFCD00002938
BTFJIXJJCSYFAL-UHFFFAOYSA-N
1-eicosanol,arachidyl alcohol,n-eicosanol,arachidic alcohol,arachic alcohol,eicosyl alcohol,eicosan-1-ol,eicosanol,1-icosanol,n-1-eicosanol
12404
CHEBI:75627
icosan-1-ol
CCCCCCCCCCCCCCCCCCCCO
629-96-9
C20H42O
298.54
MFCD00002938
BTFJIXJJCSYFAL-UHFFFAOYSA-N
1-eicosanol,arachidyl alcohol,n-eicosanol,arachidic alcohol,arachic alcohol,eicosyl alcohol,eicosan-1-ol,eicosanol,1-icosanol,n-1-eicosanol
12404
CHEBI:75627
icosan-1-ol
CCCCCCCCCCCCCCCCCCCCO
Flash Point 194°C
Quantity 5g
Melting Point 64°C to 66°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)19OH
Beilstein 01, 431
Solubility Information Solubility in water: slightly soluble
Formula Weight 298.54
Physical Form Pellets, Powder or Chunks
Percent Purity 98%
Chemical Name or Material 1-Eicosanol
Show More Show Less

EINECSNumber : 211-119-4