Alfa Aesar™ 1-Ethoxycarbonyl-4-piperidone, 99%

Manufacturer: Alfa Aesar™ A1183318

Quantity 50g

 View more versions of this product

Catalog No. AAA1183318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 29976-53-2
    Molecular Formula C8H13NO3
    Formula Weight 171.2
    MDL Number MFCD00006188
    Beilstein 640967
    Synonym N-Carboethoxy-4-piperidone; Ethyl 4-oxo-1-piperidinecarboxylate
    Chemical Name or Material 1-Ethoxycarbonyl-4-piperidone

    Specifications

    Formula Weight 171.2
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00006188
    Molecular Formula C8H13NO3
    CAS 29976-53-2
    Synonym N-Carboethoxy-4-piperidone; Ethyl 4-oxo-1-piperidinecarboxylate
    Health Hazard 1 Warning
    Beilstein 640967
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Flash Point 87°C (188°F)
    Boiling Point 95° to 98°C (1mmHg)
    Quantity 50g
    Chemical Name or Material 1-Ethoxycarbonyl-4-piperidone
    Assay 99%

    Description

    Applications
    1-Ethoxycarbonyl-4-piperidone is used in the preparation of 3-hydrazinopyridazines as antihypertensive agents as well as γ-carboline derivatives as potential serotonergic agents.

    Solubility
    Soluble in water.500 g/L at 20°C

    Notes
    Store in a cool, dry, well-ventilated area away from incompatible substances. Protect from heat. Incompatible with oxidizing agents.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit