Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethoxycarbonyl-4-piperidone, 99%, Thermo Scientific™

Click to view available options
Quantity:
10g
50g
250g
29976-53-2
C8H13NO3
171.196
MFCD00006188
LUBGFMZTGFXIIN-UHFFFAOYSA-N
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
34767
ethyl 4-oxopiperidine-1-carboxylate
CCOC(=O)N1CCC(=O)CC1
29976-53-2
C8H13NO3
171.196
MFCD00006188
LUBGFMZTGFXIIN-UHFFFAOYSA-N
n-carbethoxy-4-piperidone, ethyl 4-oxo-1-piperidinecarboxylate, n-carboethoxy-4-piperidone, 1-ethoxycarbonyl-4-piperidone, 1-carbethoxy-4-piperidone, 4-oxo-1-piperidinecarboxylic acid ethyl ester, n-ethoxycarbonyl-4-piperidone, unii-7h5jx1s259, 1-piperidinecarboxylic acid, 4-oxo-, ethyl ester, ethyl 4-oxopiperidinecarboxylate
34767
ethyl 4-oxopiperidine-1-carboxylate
CCOC(=O)N1CCC(=O)CC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Ethoxycarbonyl-4-piperidone is used in the preparation of 3-hydrazinopyridazines as antihypertensive agents as well as γ-carboline derivatives as potential serotonergic agents.

Solubility
Soluble in water.500 g/L at 20°C

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances. Protect from heat. Incompatible with oxidizing agents.
Density 1.135
Boiling Point 95°C to 98°C (1mmHg)
Flash Point 87°C (188°F)
Beilstein 640967
Refractive Index 1.475
Quantity 10g
Solubility Information Soluble in water.500g/L at 20°C
Formula Weight 171.2
Percent Purity 99%
Chemical Name or Material 1-Ethoxycarbonyl-4-piperidone

GHS H Statement
H227
Combustible liquid.

P210-P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P370+P378q-P501c

H227-H302-H315-H319

EINECSNumber : 249-984-5

TSCA : No

Recommended Storage : Ambient temperatures

RUO – Research Use Only