Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Ethoxycarbonyl-4-piperidone 97.0+%, TCI America™
Click to view available options
Quantity:
25g
500g
Specifications
Color | Yellow |
Quantity | 25g |
Boiling Point | 151°C |
Formula Weight | 171.20 |
Physical Form | Clear Liquid at 20°C |
Percent Purity | ≥97.0% (GC) |
Chemical Name or Material | 1-Ethoxycarbonyl-4-piperidone |
Safety and Handling
TSCA : No