Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Ethyl-2-pyrrolidinone, 98%

Click to view available options
Quantity:
25g
100g
500g
2687-91-4
C6H11NO
113.16
MFCD00003199
ZFPGARUNNKGOBB-UHFFFAOYSA-N
1-ethyl-2-pyrrolidinone,1-ethyl-2-pyrrolidone,n-ethyl-2-pyrrolidone,n-ethylpyrrolidone,2-pyrrolidinone, 1-ethyl,n-ethylpyrrolidinone,ethyl pyrrolidone,unii-h0229sx1cw,n-ethyl-,a-pyrrolidone,n-ethyl-2-pyrrolidinone
17595
1-ethylpyrrolidin-2-one
CCN1CCCC1=O
2687-91-4
C6H11NO
113.16
MFCD00003199
ZFPGARUNNKGOBB-UHFFFAOYSA-N
1-ethyl-2-pyrrolidinone,1-ethyl-2-pyrrolidone,n-ethyl-2-pyrrolidone,n-ethylpyrrolidone,2-pyrrolidinone, 1-ethyl,n-ethylpyrrolidinone,ethyl pyrrolidone,unii-h0229sx1cw,n-ethyl-,a-pyrrolidone,n-ethyl-2-pyrrolidinone
17595
1-ethylpyrrolidin-2-one
CCN1CCCC1=O
Quantity 500g
Flash Point 76°C (169°F)
Boiling Point 96°C to 98°C (20mmHg)
Formula Weight 113.16
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2-pyrrolidinone

GHS H Statement
H302-H315-H319-H335-H227
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA