Alfa Aesar™ 1-Ethyl-3-hydroxypyridinium bromide, 98%

Manufacturer: Alfa Aesar™ H5516914

Quantity 25g

 View more versions of this product

Catalog No. AAH5516914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 80866-84-8
    Formula Weight 204.07
    MDL Number MFCD00011995
    UN Number UN3263
    Chemical Name or Material 1-Ethyl-3-hydroxypyridinium bromide

    Specifications

    Formula Weight 204.07
    Melting Point 104° to 108°C
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    Physical Form Solid
    MDL Number MFCD00011995
    CAS 80866-84-8
    UN Number UN3263
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
    RTECS Number UU4024200
    Quantity 25g
    Color White to pale brown
    Chemical Name or Material 1-Ethyl-3-hydroxypyridinium bromide
    Assay 98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit