Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium bromide, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
65039-08-9
C6H11BrN2
191.07
MFCD03427610
GWQYPLXGJIXMMV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium bromide,3-ethyl-1-methyl-1h-imidazol-3-ium bromide,1-ethyl-3-methylmidazolium bromide,unii-xo254ye73i,1-ethyl-3-methyl-1h-imidazol-3-ium bromide,acmc-1b3a0,dsstox_cid_29300,dsstox_rid_83418,dsstox_gsid_49343,c6h11n2.br
2734235
1-ethyl-3-methylimidazol-3-ium;bromide
CCN1C=C[N+](=C1)C.[Br-]
65039-08-9
C6H11BrN2
191.07
MFCD03427610
GWQYPLXGJIXMMV-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium bromide,3-ethyl-1-methyl-1h-imidazol-3-ium bromide,1-ethyl-3-methylmidazolium bromide,unii-xo254ye73i,1-ethyl-3-methyl-1h-imidazol-3-ium bromide,acmc-1b3a0,dsstox_cid_29300,dsstox_rid_83418,dsstox_gsid_49343,c6h11n2.br
2734235
1-ethyl-3-methylimidazol-3-ium;bromide
CCN1C=C[N+](=C1)C.[Br-]
Ionic liquid
Infrared Spectrum Authentic
Color Yellow
Quantity 5g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (Argentometry)
Formula Weight 191.07
Physical Form Crystals, Crystalline Powder or Chunks
Percent Purity 98%
Chemical Name or Material 1-Ethyl-3-methylimidazolium bromide
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a

GHS Signal Word: Warning