Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
65039-09-0
C6H11ClN2
146.62
MFCD00074843
BMQZYMYBQZGEEY-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium chloride,1-ethyl-3-methyl-1h-imidazol-3-ium chloride,emim cl,unii-ph90aq1e93,1-ethyl-3-methylimidazoliumchloride,1-methyl-3-ethylimidazolium chloride,1-ethyl-3-methylimidazol-3-ium chloride,emim-cl,basionics™ st 80,dsstox_cid_21180
2734160
CHEBI:61327
1-ethyl-3-methylimidazol-3-ium;chloride
CCN1C=C[N+](=C1)C.[Cl-]
65039-09-0
C6H11ClN2
146.62
MFCD00074843
BMQZYMYBQZGEEY-UHFFFAOYSA-M
1-ethyl-3-methylimidazolium chloride,1-ethyl-3-methyl-1h-imidazol-3-ium chloride,emim cl,unii-ph90aq1e93,1-ethyl-3-methylimidazoliumchloride,1-methyl-3-ethylimidazolium chloride,1-ethyl-3-methylimidazol-3-ium chloride,emim-cl,basionics™ st 80,dsstox_cid_21180
2734160
CHEBI:61327
1-ethyl-3-methylimidazol-3-ium;chloride
CCN1C=C[N+](=C1)C.[Cl-]
Ionic liquid
Viscosity 61.2 mPa.s (80°C)
Quantity 25g
Melting Point 77.0°C to 79.0°C
Flash Point 186°C
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Argentometry)
Formula Weight 146.62
Physical Form Agglomerating Crystalline Powder
Percent Purity 97%
Chemical Name or Material 1-Ethyl-3-methylimidazolium chloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning