Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Ethyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Used in cellulose processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used in cellulose processing.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Used in cellulose processing.
Solubility
Soluble in water.
Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Specifications
Melting Point | 77°C to 80°C |
Flash Point | 186°C (367°F) |
Quantity | 5 g |
Sensitivity | Very Hygroscopic |
Solubility Information | Soluble in water. |
IUPAC Name | 1-ethyl-3-methyl-1H-imidazol-3-ium chloride |
Formula Weight | 146.62 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-Ethyl-3-methylimidazolium chloride |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | Yes |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction