Alfa Aesar™ 1-Ethyl-3-methylimidazolium methanesulfonate, 99%

Manufacturer: Alfa Aesar™ H2691318

Quantity 50g

 View more versions of this product

Catalog No. AAH2691318

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 145022-45-3
    Molecular Formula C7H14N2O3S
    Formula Weight 206.26
    MDL Number MFCD06798171
    UN Number UN3265
    Synonym ECOENG 110
    Chemical Name or Material 1-Ethyl-3-methylimidazolium methanesulfonate

    Specifications

    Formula Weight 206.26
    Molecular Formula C7H14N2O3S
    MDL Number MFCD06798171
    CAS 145022-45-3
    UN Number UN3265
    Synonym ECOENG 110
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 35°C
    Quantity 50g
    Flash Point 66°C (152°F)
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material 1-Ethyl-3-methylimidazolium methanesulfonate
    Assay 99%

    Description

    Applications
    Used as an electrolyte for the electrooxidative polymerization of pyrrole. Used for electroplating of aluminum onto steel, electroplating of chromium,„ electropolishing of steel and other metal surfaces. In polymer and biopolymer processing used as„ solvent for regenerating cellulose,„ enabling technology for manufacturing cellulose, derivatives„ dissolution and separation of lignocellulosic biomas.

    Solubility
    Soluble in water. Soluble in acetone, methanol .

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit