Alfa Aesar™ 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene, 99+%

Manufacturer: Alfa Aesar™ H2703803

Quantity 1g

 View more versions of this product

Catalog No. AAH2703803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 63221-88-5
    Molecular Formula C17H16O
    Formula Weight 236.31
    MDL Number MFCD00807428
    Synonym 1-(4-Ethylphenyl)-2-(4-methoxyphenyl)acetylene
    Chemical Name or Material 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene

    Specifications

    Formula Weight 236.31
    Molecular Formula C17H16O
    MDL Number MFCD00807428
    CAS 63221-88-5
    Synonym 1-(4-Ethylphenyl)-2-(4-methoxyphenyl)acetylene
    Boiling Point 151° to 153°C (1mmHg)
    Melting Point 35°C (N)
    Quantity 1g
    Chemical Name or Material 1-Ethyl-4-[(4-methoxyphenyl)ethynyl]benzene
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit