Alfa Aesar™ 1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene, 99+%

Manufacturer: Alfa Aesar™ H2690809

Quantity 10g

 View more versions of this product

Catalog No. AAH2690809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 117923-34-9
    Molecular Formula C22H26
    Formula Weight 290.45
    MDL Number MFCD09909569
    Synonym 4-Ethyl-4'-n-hexyldiphenylacetylene
    Chemical Name or Material 1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene

    Specifications

    Formula Weight 290.45
    Molecular Formula C22H26
    MDL Number MFCD09909569
    CAS 117923-34-9
    Synonym 4-Ethyl-4'-n-hexyldiphenylacetylene
    Boiling Point 172° to 174°C (0.5mmHg)
    Melting Point 10° to 12°C
    Quantity 10g
    Chemical Name or Material 1-Ethyl-4-[(4-n-hexylphenyl)ethynyl]benzene
    Assay ≥99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit