Alfa Aesar™ 1-Ethyl-4-[(4-n-propylphenyl)ethynyl]benzene, 99+%

Manufacturer: Alfa Aesar™ H2739803

Quantity 1g

 View more versions of this product

Catalog No. AAH2739803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102225-55-8
    Molecular Formula C19H20
    Formula Weight 248.37
    MDL Number MFCD05864410
    Synonym 4-Ethyl-4'-n-propyldiphenylacetylene
    Chemical Name or Material 1-Ethyl-4-[(4-n-propylphenyl)ethynyl]benzene

    Specifications

    Formula Weight 248.37
    Molecular Formula C19H20
    MDL Number MFCD05864410
    Synonym 4-Ethyl-4'-n-propyldiphenylacetylene
    Melting Point 49° to 51°C
    Quantity 1g
    CAS 102225-55-8
    Chemical Name or Material 1-Ethyl-4-[(4-n-propylphenyl)ethynyl]benzene
    Assay ≥99%

    Description

    Applications
    It is used as a primary and secondary intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit