Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-4-eth-1-ynylbenzene, Maybridge™

Click to view available options
Quantity:
1g
10g
25g
40307-11-7
C10H10
130.19
MFCD00173887
ZNTJVJSUNSUMPP-UHFFFAOYSA-N
4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
142425
1-ethyl-4-ethynylbenzene
CCC1=CC=C(C=C1)C#C
40307-11-7
C10H10
130.19
MFCD00173887
ZNTJVJSUNSUMPP-UHFFFAOYSA-N
4-ethylphenylacetylene, 1-ethyl-4-eth-1-ynylbenzene, benzene, 1-ethyl-4-ethynyl, 1-ethyl-4-ethynyl-benzene, ethyl-4-ethynylbenzene, pubchem10925, acmc-209jcv, 4-ethyl-1-ethynylbenzene, 1-ethynyl-4-ethylbenzene, p-ethylphenylacetylene
142425
1-ethyl-4-ethynylbenzene
CCC1=CC=C(C=C1)C#C
Quantity 25g
Density 0.93g/mL
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula HC2CC6H4CH2CH3
Formula Weight 130.19
Percent Purity 97%
Chemical Name or Material 1-Ethyl-4-eth-1-ynylbenzene

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.