Learn More
Alfa Aesar™ 1-Ethyl-4-[(p-tolyl)ethynyl]benzene, 99+%
Manufacturer:  Alfa Aesar™ H2688209
Quantity 10g
 View more versions of this product
Catalog No. AAH2688209
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
22692-80-4
220.31
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene; 1-(4-Ethylphenyl)-2-(p-tolyl)acetylene
C17H16
MFCD04038793
1-Ethyl-4-[(p-tolyl)ethynyl]benzene
Specifications
22692-80-4
220.31
1-Ethyl-4-[2-(4-methylphenyl)-1-ethynyl]benzene; 1-(4-Ethylphenyl)-2-(p-tolyl)acetylene
71° to 74°C
1-Ethyl-4-[(p-tolyl)ethynyl]benzene
C17H16
MFCD04038793
135° to 136°C (1mmHg)
10g
≥99%