Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Ethylcyclopentanol, 96%

Click to view available options
Quantity:
1g
5g
1462-96-0
C7H14O
114.188
MFCD00019281
LPCWIFPJLFCXRS-UHFFFAOYSA-N
1-ethylcyclopentanol,cyclopentanol, 1-ethyl,1-ethyl-1-cyclopentanol,acmc-1bzot,1-ethylcyclopentanol,ksc181q7f,1-ethyl-1-hydroxycyclopentane
137018
1-ethylcyclopentan-1-ol
CCC1(CCCC1)O
1462-96-0
C7H14O
114.188
MFCD00019281
LPCWIFPJLFCXRS-UHFFFAOYSA-N
1-ethylcyclopentanol,cyclopentanol, 1-ethyl,1-ethyl-1-cyclopentanol,acmc-1bzot,1-ethylcyclopentanol,ksc181q7f,1-ethyl-1-hydroxycyclopentane
137018
1-ethylcyclopentan-1-ol
CCC1(CCCC1)O
Density 0.909g/mL
Sensitivity Moisture Sensitive
Color Colorless to Yellow
Quantity 5g
Refractive Index 1.4570
UN Number UN1987
Formula Weight 114.19
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 1-Ethylcyclopentanol

GHS H Statement
H226
Flammable liquid and vapor.

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: ALCOHOLS, N.O.S.

storageNote1 : 2° to 8°C