Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Ethylimidazole-2-carboxaldehyde, 95%

Click to view available options
Quantity:
250mg
1g
111851-98-0
C6H8N2O
124.143
MFCD07380875
JINZWCUGPDJTNB-UHFFFAOYSA-N
1-ethyl-1h-imidazole-2-carbaldehyde,1h-imidazole-2-carboxaldehyde, 1-ethyl,1-ethylimidazole-2-carboxaldehyde,1-ethyl-2-formylimidazole,ksc880k6p,n-ethyl-2-imidazolecarbaldehyde,1-ethyl-2-formyl-1h-imidazole,n-ethylimidazole-2-carboxaldehyde,1-ethyl-1h-imidazole-2-carboxaldehyde,1h-imidazole-2-carboxaldehyde,1-ethyl-9ci
11062377
1-ethylimidazole-2-carbaldehyde
CCN1C=CN=C1C=O
111851-98-0
C6H8N2O
124.143
MFCD07380875
JINZWCUGPDJTNB-UHFFFAOYSA-N
1-ethyl-1h-imidazole-2-carbaldehyde,1h-imidazole-2-carboxaldehyde, 1-ethyl,1-ethylimidazole-2-carboxaldehyde,1-ethyl-2-formylimidazole,ksc880k6p,n-ethyl-2-imidazolecarbaldehyde,1-ethyl-2-formyl-1h-imidazole,n-ethylimidazole-2-carboxaldehyde,1-ethyl-1h-imidazole-2-carboxaldehyde,1h-imidazole-2-carboxaldehyde,1-ethyl-9ci
11062377
1-ethylimidazole-2-carbaldehyde
CCN1C=CN=C1C=O
Quantity 250mg
Beilstein 3538106
Formula Weight 124.14
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 1-Ethylimidazole-2-carboxaldehyde

storageNote1 : -30° to -10°C