Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethylpiperidine, 99%, ACROS Organics™

Click to view available options
Quantity:
500mL
2.5L
Packaging:
Glass bottle
766-09-6
C7H15N
113.2
MFCD00006507
HTLZVHNRZJPSMI-UHFFFAOYSA-N
n-ethylpiperidine,piperidine, 1-ethyl,n-aethylpiperidin,n-ethyl piperidine,ethylpiperidine,n-aethylpiperidin german,1-ethyl-piperidine,n-ethyl-piperidine,1-ethyl piperidine,1-ethylpiperidene
13007
CHEBI:39017
1-ethylpiperidine
CCN1CCCCC1
766-09-6
C7H15N
113.2
MFCD00006507
HTLZVHNRZJPSMI-UHFFFAOYSA-N
n-ethylpiperidine,piperidine, 1-ethyl,n-aethylpiperidin,n-ethyl piperidine,ethylpiperidine,n-aethylpiperidin german,1-ethyl-piperidine,n-ethyl-piperidine,1-ethyl piperidine,1-ethylpiperidene
13007
CHEBI:39017
1-ethylpiperidine
CCN1CCCCC1
Quantity 2.5L
Flash Point 17°C
Density 0.8240g/mL
Color Colorless to Yellow
Packaging Glass bottle
Specific Gravity 0.824
Refractive Index 1.4430 to 1.4450
Melting Point -20.0°C
Infrared Spectrum Authentic
Boiling Point 129.0°C to 131.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20,17
Solubility Information Solubility in water: 50g/L (20°C)
Formula Weight 113.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1-Ethylpiperidine, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 212-161-6

RTECSNumber : TN0250000

TSCA : TSCA