Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethylpyridinium chloride, 98%, Acros Organics

Click to view available options
Quantity:
25g
2294-38-4
C7H10ClN
143.614
MFCD03093293
AMFMJCAPWCXUEI-UHFFFAOYSA-M
1-ethylpyridinium chloride,ethylpyridinium chloride,1-ethylpyridin-1-ium chloride,unii-38bql5563g,pyridinium, 1-ethyl-, chloride,n-ethylpyridinium chloride,pyridinium, 1-ethyl-, chloride 1:1,1-ethylpyridiniumchloride,pyridine ethochloride,ethylpyridiniumchloride
75299
1-ethylpyridin-1-ium;chloride
CC[N+]1=CC=CC=C1.[Cl-]
2294-38-4
C7H10ClN
143.614
MFCD03093293
AMFMJCAPWCXUEI-UHFFFAOYSA-M
1-ethylpyridinium chloride,ethylpyridinium chloride,1-ethylpyridin-1-ium chloride,unii-38bql5563g,pyridinium, 1-ethyl-, chloride,n-ethylpyridinium chloride,pyridinium, 1-ethyl-, chloride 1:1,1-ethylpyridiniumchloride,pyridine ethochloride,ethylpyridiniumchloride
75299
1-ethylpyridin-1-ium;chloride
CC[N+]1=CC=CC=C1.[Cl-]
Ionic liquid
Quantity 25g
Color Beige
Infrared Spectrum Authentic
Water 1.5% max.
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 143.62
Physical Form Crystals and Agglomerates
Percent Purity ≥97.5%
Chemical Name or Material 1-Ethylpyridinium chloride
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.