Alfa Aesar™ 1-Ethynylcyclohexylamine, 98%

Manufacturer: Alfa Aesar™ H5351406

Quantity 5g

 View more versions of this product

Catalog No. AAH5351406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 30389-18-5
    Molecular Formula C8H13N
    Formula Weight 123.2
    MDL Number MFCD00001489
    UN Number UN2734
    Chemical Name or Material 1-Ethynylcyclohexylamine

    Specifications

    Formula Weight 123.2
    TSCA TSCA
    Molecular Formula C8H13N
    MDL Number MFCD00001489
    CAS 30389-18-5
    UN Number UN2734
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
    Flash Point 42°C (108°F)
    Boiling Point 65° to 66°C (20mmHg)
    Quantity 5g
    Chemical Name or Material 1-Ethynylcyclohexylamine
    Assay 98%

    Description

    Applications
    1-Ethynylcyclohexylamine was used in the synthesis of bicyclic and tetracyclic compounds. It was also used as a substrate in the reaction of a series of alkynes with mercury(II)acetate.

    Notes
    Air sensitive. Incompatible with oxidizing agents and acids.

    GHS H Statement
    H314-H318-H226
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapor.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit