Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Ethynylcyclopentanol, 98+%

Click to view available options
Quantity:
5g
25g
100g
17356-19-3
C7H10O
110.156
MFCD00001365
LQMDOONLLAJAPZ-UHFFFAOYSA-N
1-ethynylcyclopentanol,cyclopentanol, 1-ethynyl,1-ethynyl-1-cyclopentanol,cyclopentyl ethynyl carbinol,1-ethynyl-cyclopentan-1-ol,acmc-20abvz,1-ethinylcyclopentanol,1-ethynyl-cyclopentanol,cyclopentanol, ethynyl,1-ethynylcyclopentanol ?
87074
1-ethynylcyclopentan-1-ol
C#CC1(CCCC1)O
17356-19-3
C7H10O
110.156
MFCD00001365
LQMDOONLLAJAPZ-UHFFFAOYSA-N
1-ethynylcyclopentanol,cyclopentanol, 1-ethynyl,1-ethynyl-1-cyclopentanol,cyclopentyl ethynyl carbinol,1-ethynyl-cyclopentan-1-ol,acmc-20abvz,1-ethinylcyclopentanol,1-ethynyl-cyclopentanol,cyclopentanol, ethynyl,1-ethynylcyclopentanol ?
87074
1-ethynylcyclopentan-1-ol
C#CC1(CCCC1)O
Applications
1-Ethynylcyclopentanol is used in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Boiling Point 156°C to 159°C
Flash Point 48°C (118°F)
Melting Point 23°C to 27°C
Quantity 5g
Odor Irritating
UN Number UN1325
Beilstein 1924167
Formula Weight 110.16
Percent Purity ≥98%
Chemical Name or Material 1-Ethynylcyclopentanol

GHS H Statement
H228-H302-H319-H335
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280f-P305+P351+P338
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA