Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-Fluoro-2,4-dimethoxybenzene, 97%

Click to view available options
Quantity:
250mg
1g
5g
17715-70-7
C8H9FO2
156.156
MFCD00042265
QLJNEPOEZGFNEA-UHFFFAOYSA-N
2,4-dimethoxy-1-fluorobenzene,2,4-dimethoxyfluorobenzene,1,3-dimethoxy-4-fluorobenzene,4-fluoro-3-methoxyanisole,benzene, 1-fluoro-2,4-dimethoxy,2,4-dimethoxy-1-fluoro-benzene,acmc-1c4p9,1-fluoro-2,4-dimethoxy-benzene,qljnepoezgfnea-uhfffaoysa,benzene,1-fluoro-2,4-dimethoxy
2758413
1-fluoro-2,4-dimethoxybenzene
COC1=CC(=C(C=C1)F)OC
17715-70-7
C8H9FO2
156.156
MFCD00042265
QLJNEPOEZGFNEA-UHFFFAOYSA-N
2,4-dimethoxy-1-fluorobenzene,2,4-dimethoxyfluorobenzene,1,3-dimethoxy-4-fluorobenzene,4-fluoro-3-methoxyanisole,benzene, 1-fluoro-2,4-dimethoxy,2,4-dimethoxy-1-fluoro-benzene,acmc-1c4p9,1-fluoro-2,4-dimethoxy-benzene,qljnepoezgfnea-uhfffaoysa,benzene,1-fluoro-2,4-dimethoxy
2758413
1-fluoro-2,4-dimethoxybenzene
COC1=CC(=C(C=C1)F)OC
Flash Point 105°C (221°F)
Density 1.188 g/mL
Color Yellow
Quantity 5g
Formula Weight 156.15
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 1-Fluoro-2,4-dimethoxybenzene

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 605-794-8

TSCA : No

Recommended Storage : Ambient temperatures